missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl 4-Chlorophenyl Sulfone 95.0+%, TCI America™

Catalog No. C13105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13105G Supplier TCI America Supplier No. C13105G
Only null left

Chemical Identifiers

CAS 16191-84-7
Molecular Formula C8H8Cl2O2S
Molecular Weight (g/mol) 239.11
MDL Number MFCD00018966
InChI Key KXQHTLXSDBXWNB-UHFFFAOYSA-N
PubChem CID 27745
IUPAC Name 1-chloro-4-(2-chloroethanesulfonyl)benzene
SMILES ClCCS(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.11
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethyl 4-Chlorophenyl Sulfone
RTECSNumber WR5350000
TSCA No