missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorocyclohexanol 95.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 84°C |
| Quantity | 5 g |
| Formula Weight | 134.60 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 2-Chlorocyclohexanol |
Safety and Handling
| TSCA | No |