accessibility menu, dialog, popup

UserName

OHCl

CAS RN 1561-86-0

IUPAC Name: 2-chlorocyclohexan-1-ol
Synonyms: 2-chlorocyclohexan-1-ol
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: NYEWDMNOXFGGDX-UHFFFAOYNA-N
SMILES: OC1CCCCC1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results