missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Chloride 99.0+%, TCI America™

Catalog No. D042025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D042025G 25 g
D0420500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D042025G Supplier TCI America Supplier No. D042025G

Chemical Identifiers

CAS 611-19-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000893
InChI Key BASMANVIUSSIIM-UHFFFAOYSA-N
Synonym 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd
PubChem CID 11906
IUPAC Name 1-chloro-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Cl

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 g
UN Number 2235
Formula Weight 161.03
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzyl Chloride
EINECSNumber (3)-0091&(3)-0078
RTECSNumber CZ0195000
TSCA Yes
Recommended Storage Refrigerator