Learn More
2-Chlorobenzyl chloride, 98+%
CAS: 611-19-8 | C7H6Cl2 | 161.025 g/mol
Supplier: Thermo Scientific Chemicals A119000E
Description
2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.
Solubility
Not miscible in water.
Notes
Store away from strong bases and oxidizing agents. Keep away from heat. Incompatible with Alcohols, Amines, Metals.
Specifications
| 2-Chlorobenzyl chloride | |
| -13°C | |
| 213°C to 214°C | |
| Pungent | |
| 1.559 | |
| 2500 g | |
| 471700 | |
| Not miscible in water. | |
| C1=CC=C(C(=C1)CCl)Cl | |
| 161.025 | |
| 161.03 |
| 611-19-8 | |
| 1.272 | |
| 109°C (228°F) | |
| C7H6Cl2 | |
| MFCD00000893 | |
| UN2235 | |
| 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd | |
| BASMANVIUSSIIM-UHFFFAOYSA-N | |
| 1-chloro-2-(chloromethyl)benzene | |
| 11906 | |
| ≥98% |
Chemical Identifiers
| 611-19-8 | |
| 161.025 | |
| BASMANVIUSSIIM-UHFFFAOYSA-N | |
| 11906 | |
| C1=CC=C(C(=C1)CCl)Cl |
| C7H6Cl2 | |
| MFCD00000893 | |
| 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd | |
| 1-chloro-2-(chloromethyl)benzene |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302+H312+H332-H314-H317-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROBENZYL CHLORIDES, LIQUID
EINECSNumber : 210-258-8
RTECSNumber : CZ0195000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only