missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl chloride, 98+%

Catalog No. AAA1190036
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1190036 500 g
AAA119000E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1190036 Supplier Thermo Scientific Chemicals Supplier No. A1190036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-19-8 | C7H6Cl2 | 161.025 g/mol

2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzyl chloride was used as a sensory irritant to evaluate the characteristic modifications of the normal breathing pattern of exposed mice. It is used as an intermediate for organic chemicals, pharmaceuticals, and dyes.

Solubility
Not miscible in water.

Notes
Store away from strong bases and oxidizing agents. Keep away from heat. Incompatible with Alcohols, Amines, Metals.

Chemical Identifiers

CAS 611-19-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000893
InChI Key BASMANVIUSSIIM-UHFFFAOYSA-N
Synonym 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd
PubChem CID 11906
IUPAC Name 1-chloro-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Cl

Specifications

Melting Point -13°C
Density 1.272
Boiling Point 213°C to 214°C
Flash Point 109°C (228°F)
Odor Pungent
Refractive Index 1.559
Quantity 500 g
UN Number UN2235
Beilstein 471700
Solubility Information Not miscible in water.
Formula Weight 161.03
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzyl chloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H317-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROBENZYL CHLORIDES, LIQUID
EINECSNumber 210-258-8
RTECSNumber CZ0195000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only