missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl bromide, 97%

Catalog No. AAA1212914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1212914 25 g
AAA1212906 5 g
AAA1212922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1212914 Supplier Thermo Scientific Chemicals Supplier No. A1212914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-17-6 | C7H6BrCl | 205.479 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-17-6
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00000566
InChI Key PURSZYWBIQIANP-UHFFFAOYSA-N
Synonym 2-chlorobenzyl bromide, 1-bromomethyl-2-chlorobenzene, o-chlorobenzyl bromide, 2-chlorobenzylbromide, benzene, 1-bromomethyl-2-chloro, alpha-bromo-2-chlorotoluene, unii-cev23h8uv4, 2chlorobenzyl bromide, alpha-bromo-o-chlorotoluene, cev23h8uv4
PubChem CID 11905
IUPAC Name 1-(bromomethyl)-2-chlorobenzene
SMILES C1=CC=C(C(=C1)CBr)Cl

Specifications

Density 1.583
Boiling Point 103°C to 104°C (10 mmHg)
Flash Point 108°C (226°F)
Odor Pungent
Refractive Index 1.592
Quantity 25 g
UN Number UN3265
Beilstein 386264
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 210-257-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only