missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoxazole, 98%, Thermo Scientific™

Chemical Identifiers

CAS 615-18-9
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005766
InChI Key BBVQDWDBTWSGHQ-UHFFFAOYSA-N
Synonym 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
PubChem CID 11986
IUPAC Name 2-chloro-1,3-benzoxazole
SMILES ClC1=NC2=CC=CC=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC404530100
SDS View Documents
Thermo Scientific Chemicals
404530100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC404530500
SDS View Documents
Thermo Scientific Chemicals
404530500
50g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

615-18-9
153.57
BBVQDWDBTWSGHQ-UHFFFAOYSA-N
11986
ClC1=NC2=CC=CC=C2O1
C7H4ClNO
MFCD00005766
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
2-chloro-1,3-benzoxazole

Specifications

615-18-9
100.0
200.0°C
Authentic
Glass bottle
1.5655 to 1.5675
10g
2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
ClC1=NC2=CC=CC=C2O1
153.57
153.57
2-Chlorobenzoxazole, 99%
98.5
1.3500g/mL
85°C
97.5% min. (GC)
C7H4ClNO
MFCD00005766
1.35
BBVQDWDBTWSGHQ-UHFFFAOYSA-N
2-chloro-1,3-benzoxazole
11986
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 210-414-5

RTECSNumber : DM4680000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only