missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoxazole, 98%

Catalog No. AC404530100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404530100 10 g
AC404530500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404530100 Supplier Thermo Scientific Chemicals Supplier No. 404530100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 615-18-9 | C7H4ClNO | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-18-9
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005766
InChI Key BBVQDWDBTWSGHQ-UHFFFAOYSA-N
Synonym 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
PubChem CID 11986
IUPAC Name 2-chloro-1,3-benzoxazole
SMILES ClC1=NC2=CC=CC=C2O1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 7.0°C
Color White
Density 1.3500g/mL
Boiling Point 200.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5655 to 1.5675
Quantity 10 g
Specific Gravity 1.35
Formula Weight 153.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chlorobenzoxazole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 210-414-5
RTECSNumber DM4680000
TSCA TSCA

RUO – Research Use Only