missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzotrifluoride 98.0+%, TCI America™

Numéro de catalogue. p-7124348
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C030325G 25 g
C0303500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C030325G Fournisseur TCI America Code fournisseur C030325G
Il en reste null

Identifiants chimiques

CAS 88-16-4
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000561
InChI Key DGRVQOKCSKDWIH-UHFFFAOYSA-N
Synonym 2-chlorobenzotrifluoride, 1-chloro-2-trifluoromethyl benzene, o-chlorobenzotrifluoride, chlorobenzotrifluoride, benzene, 1-chloro-2-trifluoromethyl, 2-chloro trifluoromethyl benzene, 2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-benzotrifluoride, o-trifluoromethyl chlorobenzene, o-trifluoromethyl phenyl chloride
PubChem CID 6921
IUPAC Name 1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Cl

Spécifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 2234
Formula Weight 180.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzotrifluoride
EINECSNumber (3)-0053
RTECSNumber XS9144000
TSCA Yes