missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzotrifluoride, 99%

Catalog No. p-7080979
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1075322 100 g
AAA1075336 500 g
AAA107530E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1075322 Supplier Thermo Scientific Chemicals Supplier No. A1075322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-16-4 | C7H4ClF3 | 180.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-16-4
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000561
InChI Key DGRVQOKCSKDWIH-UHFFFAOYSA-N
Synonym 2-chlorobenzotrifluoride, 1-chloro-2-trifluoromethyl benzene, o-chlorobenzotrifluoride, chlorobenzotrifluoride, benzene, 1-chloro-2-trifluoromethyl, 2-chloro trifluoromethyl benzene, 2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-benzotrifluoride, o-trifluoromethyl chlorobenzene, o-trifluoromethyl phenyl chloride
PubChem CID 6921
IUPAC Name 1-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)Cl

Specifications

Melting Point -6°C
Density 1.369
Boiling Point 151°C to 153°C
Flash Point 58°C (136°F)
Refractive Index 1.456
Quantity 100 g
UN Number UN2234
Beilstein 510993
Formula Weight 180.56
Percent Purity 99%
Chemical Name or Material 2-Chlorobenzotrifluoride
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES
EINECSNumber 201-805-1
RTECSNumber XS9141000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only