missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzothiazole 98.0+%, TCI America™
Supplier: TCI America C0332250G
Specifications
| 2-Chlorobenzothiazole | |
| White-Yellow | |
| C7H4ClNS | |
| 250 g | |
| BSQLQMLFTHJVKS-UHFFFAOYSA-N | |
| 2-chloro-1,3-benzothiazole | |
| 11987 | |
| ≥98.0% (GC) |
| 615-20-3 | |
| 248°C | |
| MFCD00005776 | |
| 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole | |
| C1=CC=C2C(=C1)N=C(S2)Cl | |
| 169.626 | |
| 169.63 | |
| Crystalline Lumps |
Chemical Identifiers
| 615-20-3 | |
| 169.626 | |
| BSQLQMLFTHJVKS-UHFFFAOYSA-N | |
| 11987 | |
| C1=CC=C2C(=C1)N=C(S2)Cl |
| C7H4ClNS | |
| MFCD00005776 | |
| 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole | |
| 2-chloro-1,3-benzothiazole |
Safety and Handling
EINECSNumber : (5)-6513
RTECSNumber : DL3150000
TSCA : Yes