missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzothiazole 98.0+%, TCI America™

Catalog No. C033225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C033225G 25 g
C0332250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C033225G Supplier TCI America Supplier No. C033225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 615-20-3
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00005776
InChI Key BSQLQMLFTHJVKS-UHFFFAOYSA-N
Synonym 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
PubChem CID 11987
IUPAC Name 2-chloro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Cl

Specifications

Color White-Yellow
Boiling Point 248°C
Quantity 25 g
Formula Weight 169.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chlorobenzothiazole
EINECSNumber (5)-6513
RTECSNumber DL3150000
TSCA Yes