missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzothiazole, 98%

Catalog No. AAA1776206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1776206 5 g
AAA1776214 25 g
AAA1776222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1776206 Supplier Thermo Scientific Chemicals Supplier No. A1776206

CAS: 615-20-3 | C7H4ClNS | 169.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 615-20-3
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00005776
InChI Key BSQLQMLFTHJVKS-UHFFFAOYSA-N
Synonym 2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
PubChem CID 11987
IUPAC Name 2-chloro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Cl

Specifications

Melting Point 30°C to 33°C
Density 1.37
Boiling Point 141°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.637
Quantity 5 g
Beilstein 116316
Formula Weight 169.63
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-415-0
RTECSNumber DL3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only