missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzophenone 98.0+%, TCI America™

$80.85 - $660.33

Chemical Identifiers

CAS 3-8-5162
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000558
InChI Key VMHYWKBKHMYRNF-UHFFFAOYSA-N
Synonym 2-chlorobenzophenone, 2-chlorophenyl phenyl methanone, methanone, 2-chlorophenyl phenyl, o-chlorobenzophenone, 2-chlorophenyl phenylmethanone, benzophenone, 2-chloro, 2-chlorophenyl-phenylmethanone, 2-chlorobenyl phenyl ketone, 2-chloro-phenyl-phenyl-methanone, benzophenone, 2-chloro-6ci,7ci,8ci
PubChem CID 78838
IUPAC Name (2-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
C115025G
View Documents
TCI America
C115025G
25 g
Each for $80.85
Only null left
 
C1150100G
View Documents
TCI America
C1150100G
100 g
Each for $207.92
Only null left
 
C1150500G
View Documents
TCI America
C1150500G
500 g
Each for $660.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3-8-5162
216.66
VMHYWKBKHMYRNF-UHFFFAOYSA-N
78838
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
C13H9ClO
MFCD00000558
2-chlorobenzophenone, 2-chlorophenyl phenyl methanone, methanone, 2-chlorophenyl phenyl, o-chlorobenzophenone, 2-chlorophenyl phenylmethanone, benzophenone, 2-chloro, 2-chlorophenyl-phenylmethanone, 2-chlorobenyl phenyl ketone, 2-chloro-phenyl-phenyl-methanone, benzophenone, 2-chloro-6ci,7ci,8ci
(2-chlorophenyl)-phenylmethanone

Specifications

3-8-5162
White-Yellow
C13H9ClO
25 g
VMHYWKBKHMYRNF-UHFFFAOYSA-N
(2-chlorophenyl)-phenylmethanone
78838
≥98.0% (GC)
2-Chlorobenzophenone
46°C
180°C
MFCD00000558
2-chlorobenzophenone, 2-chlorophenyl phenyl methanone, methanone, 2-chlorophenyl phenyl, o-chlorobenzophenone, 2-chlorophenyl phenylmethanone, benzophenone, 2-chloro, 2-chlorophenyl-phenylmethanone, 2-chlorobenyl phenyl ketone, 2-chloro-phenyl-phenyl-methanone, benzophenone, 2-chloro-6ci,7ci,8ci
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl
216.66
216.66
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : PC4945633

TSCA : Yes

SDS
Documents

Documents