missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzophenone 98.0+%, TCI America™

Catalog No. C115025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C115025G 25 g
C1150100G 100 g
C1150500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C115025G Supplier TCI America Supplier No. C115025G

Chemical Identifiers

CAS 3-8-5162
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000558
InChI Key VMHYWKBKHMYRNF-UHFFFAOYSA-N
Synonym 2-chlorobenzophenone, 2-chlorophenyl phenyl methanone, methanone, 2-chlorophenyl phenyl, o-chlorobenzophenone, 2-chlorophenyl phenylmethanone, benzophenone, 2-chloro, 2-chlorophenyl-phenylmethanone, 2-chlorobenyl phenyl ketone, 2-chloro-phenyl-phenyl-methanone, benzophenone, 2-chloro-6ci,7ci,8ci
PubChem CID 78838
IUPAC Name (2-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2Cl

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 180°C
Quantity 25 g
Formula Weight 216.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzophenone
RTECSNumber PC4945633
TSCA Yes