missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzimidazole 98.0+%, TCI America™

Catalog No. C17195G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17195G Supplier TCI America Supplier No. C17195G

Chemical Identifiers

CAS 4857-06-1
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00051944
InChI Key AYPSHJCKSDNETA-UHFFFAOYSA-N
Synonym 2-chlorobenzimidazole, 2-chloro-1h-benzo d imidazole, 1h-benzimidazole, 2-chloro, 2-chloro-1h-benzoimidazole, 2-chloro-1h-1,3-benzodiazole, benzimidazole, 2-chloro, 2-chloro-1h-1,3-benzimidazole, 2-chlorobenzoimidazole, pubchem7598, 2-chloro benzimidazole
PubChem CID 78572
IUPAC Name 2-chloro-1H-1,3-benzodiazole
SMILES ClC1=NC2=CC=CC=C2N1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 152.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzimidazole