missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzimidazole, 97%

Catalog No. AAA1219314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1219314 25 g
AAA1219306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1219314 Supplier Thermo Scientific Chemicals Supplier No. A1219314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol

2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.

Chemical Identifiers

CAS 4857-06-1
MDL Number MFCD00051944

Specifications

Melting Point ∼180°C to 185°C
Molecular Formula C7H5ClN2
Quantity 25 g
Beilstein 116526
Synonym 2-chlorobenzimidazole, 2-chloro-1h-benzo d imidazole, 1h-benzimidazole, 2-chloro, 2-chloro-1h-benzoimidazole, 2-chloro-1h-1,3-benzodiazole, benzimidazole, 2-chloro, 2-chloro-1h-1,3-benzimidazole, 2-chlorobenzoimidazole, pubchem7598, 2-chloro benzimidazole
Solubility Information Insoluble in water.
InChI Key AYPSHJCKSDNETA-UHFFFAOYSA-N
SMILES ClC1=NC2=CC=CC=C2N1
IUPAC Name 2-chloro-1H-benzimidazole
Molecular Weight (g/mol) 152.58
PubChem CID 78572
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzimidazole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-453-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only