missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzenethiol 97.0+%, TCI America™
Supplier: TCI America C197525G
Specifications
| 2-Chlorobenzenethiol | |
| Yellow | |
| C6H5ClS | |
| 1760 | |
| PWOBDMNCYMQTCE-UHFFFAOYSA-N | |
| 2-chlorobenzenethiol | |
| 80599 | |
| ≥97.0% (GC) |
| 6320-03-2 | |
| 206°C | |
| 25 g | |
| 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol | |
| C1=CC=C(C(=C1)S)Cl | |
| 144.62 | |
| 144.62 | |
| Liquid |
Chemical Identifiers
| 6320-03-2 | |
| 144.62 | |
| 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol | |
| 2-chlorobenzenethiol |
| C6H5ClS | |
| PWOBDMNCYMQTCE-UHFFFAOYSA-N | |
| 80599 | |
| C1=CC=C(C(=C1)S)Cl |
Safety and Handling
TSCA : Yes