missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenethiol 97.0+%, TCI America™

Catalog No. C19755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19755G 5 g
C197525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19755G Supplier TCI America Supplier No. C19755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6320-03-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
PubChem CID 80599
IUPAC Name 2-chlorobenzenethiol
SMILES C1=CC=C(C(=C1)S)Cl

Specifications

Color Yellow
Boiling Point 206°C
Quantity 5 g
UN Number 1760
Formula Weight 144.62
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzenethiol