missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzamide 98.0+%, TCI America™

Catalog No. C056425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C056425G Supplier TCI America Supplier No. C056425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-66-5
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007973
InChI Key RBGDLYUEXLWQBZ-UHFFFAOYSA-N
Synonym o-chlorobenzamide, benzamide, 2-chloro, benzamide, o-chloro, o-chloro-benzamide, unii-ke1m5i9i32, chlorobenzamide, pubchem3617, acmc-1aupu, 2-chlorobenzamide, dsstox_cid_30850
PubChem CID 69111
IUPAC Name 2-chlorobenzamide
SMILES NC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point 142°C
Color White
Quantity 25 g
Formula Weight 155.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzamide
EINECSNumber (3)-1430
RTECSNumber CV2443737
TSCA Yes