missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzamide, 98%

Catalog No. AAA1570614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1570614 25 g
AAA1570622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1570614 Supplier Thermo Scientific Chemicals Supplier No. A1570614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 609-66-5 | C7H6ClNO | 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-66-5
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007973
InChI Key RBGDLYUEXLWQBZ-UHFFFAOYSA-N
Synonym o-chlorobenzamide, benzamide, 2-chloro, benzamide, o-chloro, o-chloro-benzamide, unii-ke1m5i9i32, chlorobenzamide, pubchem3617, acmc-1aupu, 2-chlorobenzamide, dsstox_cid_30850
PubChem CID 69111
IUPAC Name 2-chlorobenzamide
SMILES NC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point 140°C to 144°C
Quantity 25 g
Beilstein 508510
Formula Weight 155.58
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzamide
Hazard Category H302
Hazard Statement GHS H Statement
H330-H302-H315-H319-H335
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-195-6
RTECSNumber CV2443737
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only