missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroadenosine

Catalog No. AAJ63810MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63810MD 250 mg
AAJ6381003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63810MD Supplier Thermo Scientific Chemicals Supplier No. J63810MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 146-77-0 | C10H12ClN5O4 | 301.69 g/mol

2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

Solubility
Soluble in water.

Notes
protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 146-77-0
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005734
InChI Key BIXYYZIIJIXVFW-SBTUYZBDNA-N
Synonym 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado
PubChem CID 8974
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1

Specifications

Color White
Quantity 250 mg
Beilstein 43957
Solubility Information Soluble in water.
Formula Weight 301.69
Physical Form Powder
Chemical Name or Material 2-Chloroadenosine
EINECSNumber 205-678-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only