missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroadenosine Hydrate 98.0+%, TCI America™
$48.46 - $281.48
Chemical Identifiers
| CAS | 146-77-0 |
|---|---|
| Molecular Formula | C10H12ClN5O4 |
| Molecular Weight (g/mol) | 301.69 |
| MDL Number | MFCD00005734 |
| InChI Key | BIXYYZIIJIXVFW-SBTUYZBDNA-N |
| Synonym | 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado |
| PubChem CID | 8974 |
| IUPAC Name | (2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2192100MG
|
TCI America
C2192100MG |
100 mg |
Each for $48.46
|
|
|||||
|
C21921G
|
TCI America
C21921G |
1 g |
Each for $281.48
|
|
|||||
Chemical Identifiers
| 146-77-0 | |
| 301.69 | |
| BIXYYZIIJIXVFW-SBTUYZBDNA-N | |
| 8974 | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1 |
| C10H12ClN5O4 | |
| MFCD00005734 | |
| 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado | |
| (2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
Specifications
| 146-77-0 | |
| White-Yellow | |
| MFCD00005734 | |
| 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado | |
| NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1 | |
| 301.69 | |
| 301.69 | |
| Crystalline Powder |
| 162°C | |
| C10H12ClN5O4 | |
| 100 mg | |
| BIXYYZIIJIXVFW-SBTUYZBDNA-N | |
| (2R,3R,4S,5R)-2-(6-amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol | |
| 8974 | |
| ≥98.0% (HPLC,T) | |
| 2-Chloroadenosine Hydrate |
Safety and Handling
RTECSNumber : AU7357550
TSCA : No