missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroacetamide 98.0+%, TCI America™
$48.13 - $103.97
Chemical Identifiers
| CAS | 79-07-2 |
|---|---|
| Molecular Formula | C2H4ClNO |
| Molecular Weight (g/mol) | 93.51 |
| MDL Number | MFCD00008027 |
| InChI Key | VXIVSQZSERGHQP-UHFFFAOYSA-N |
| Synonym | chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide |
| PubChem CID | 6580 |
| IUPAC Name | 2-chloroacetamide |
| SMILES | NC(=O)CCl |
Chemical Identifiers
| 79-07-2 | |
| 93.51 | |
| VXIVSQZSERGHQP-UHFFFAOYSA-N | |
| 6580 | |
| NC(=O)CCl |
| C2H4ClNO | |
| MFCD00008027 | |
| chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
| 2-chloroacetamide |
Specifications
| 79-07-2 | |
| White | |
| C2H4ClNO | |
| 25 g | |
| chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide | |
| NC(=O)CCl | |
| 93.51 | |
| 93.51 | |
| Crystalline Powder |
| 120°C | |
| 225°C | |
| MFCD00008027 | |
| 2811 | |
| VXIVSQZSERGHQP-UHFFFAOYSA-N | |
| 2-chloroacetamide | |
| 6580 | |
| ≥98.0% (GC) | |
| 2-Chloroacetamide |
Safety and Handling
EINECSNumber : (2)-1148&(2)-2642
RTECSNumber : AB5075000
TSCA : Yes