missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamide, 98+%

CAS: 79-07-2 | C2H4ClNO | 93.51 g/mol

Supplier:  Thermo Scientific Chemicals A152380I

 View more versions of this product

Catalog No. AAA152380I

Only null left
Explore available promotions

Description

Description

2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.

Solubility
Soluble in water(90g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

2-Chloroacetamide
116°C to 119°C
170°C (338°F)
C2H4ClNO
5000 g
878191
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
VXIVSQZSERGHQP-UHFFFAOYSA-N
2-chloroacetamide
6580
≥98%
79-07-2
220°C (decomposition)
Weak
MFCD00008027
UN2811
14,2110
Soluble in water(90g/L).
NC(=O)CCl
93.51
93.51

Chemical Identifiers

79-07-2
93.51
VXIVSQZSERGHQP-UHFFFAOYSA-N
6580
NC(=O)CCl
C2H4ClNO
MFCD00008027
chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
2-chloroacetamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H361-H317
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P310-P302+P352-P308+P313-P330-P333+P313-P363-P501c

H301-H317-H361f-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 201-174-2

RTECSNumber : AB5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only