missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-p-cresol 97.0+%, TCI America™

$54.06 - $205.05

Chemical Identifiers

CAS 6640-27-3
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
InChI Key AQJFATAFTQCRGC-UHFFFAOYSA-N
Synonym 2-Chloro-4-methylphenol
PubChem CID 14851
ChEBI CHEBI:79732
IUPAC Name 2-chloro-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C20055G
View Documents
TCI America
C20055G
5 g N/A
Only null left
 
C200525G
View Documents
TCI America
C200525G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6640-27-3
142.582
2-Chloro-4-methylphenol
CHEBI:79732
CC1=CC(=C(C=C1)O)Cl
C7H7ClO
AQJFATAFTQCRGC-UHFFFAOYSA-N
14851
2-chloro-4-methylphenol

Specifications

6640-27-3
196°C
5 g
2-Chloro-4-methylphenol
CC1=CC(=C(C=C1)O)Cl
142.582
CHEBI:79732
≥97.0% (GC)
2-Chloro-p-cresol
Yellow
C7H7ClO
2669
AQJFATAFTQCRGC-UHFFFAOYSA-N
2-chloro-4-methylphenol
14851
142.58
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0900

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'