missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-phenylbenzamide, 97%, Thermo Scientific Chemicals

Catalog No. p-7053322
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59623MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH59623MD Supplier Thermo Scientific Chemicals Supplier No. H59623MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6833-13-2 | Molecular Formula: C13H10ClNO | Molecular Weight: 231.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6833-13-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00016336
InChI Key AXQVIWHAEYLGLO-UHFFFAOYSA-N
Synonym 2-chlorobenzanilide, o-chlorobenzanilide, 2-chlorobenzoanilide, benzamide, 2-chloro-n-phenyl, 2-chlorobenzoic acid anilide, benzanilide, 2-chloro, n-phenyl-2-chlorobenzamide, 2-chloro-n-phenyl benzamide, 2-chloro-n-phenyl-benzamide, 2-chlorophenyl-n-benzamide
PubChem CID 23265
IUPAC Name 2-chloro-N-phenylbenzamide
SMILES ClC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 231.68
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-N-phenylbenzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only