missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylaniline 98.0+%, TCI America™

Catalog No. p-7125839
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31211G 1 g
C31215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31211G Supplier TCI America Supplier No. C31211G

Chemical Identifiers

CAS 932-32-1
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00045170
InChI Key WGNNILPYHCKCFF-UHFFFAOYSA-N
Synonym n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline
PubChem CID 136736
IUPAC Name 2-chloro-N-methylaniline
SMILES CNC1=CC=CC=C1Cl

Specifications

Boiling Point 218°C
Quantity 1 g
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-N-methylaniline