missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylaniline, 97%

Catalog No. p-7021840
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1434303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1434303 Supplier Thermo Scientific Chemicals Supplier No. A1434303

CAS: 932-32-1 | C7H8ClN | 141.598 g/mol

2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson's reagent and solvent tetrahydrofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N-methylaniline is used to produce 1-methyl-2-thioacetylpyrrole in the presence of reagent Lawesson′s reagent and solvent tetrahydrofuran.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 932-32-1
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00045170
InChI Key WGNNILPYHCKCFF-UHFFFAOYSA-N
Synonym n-methyl-2-chloroaniline, benzenamine, 2-chloro-n-methyl, n1-methyl-2-chloroaniline, 2-chloro-n-methylbenzenamine, o-chlormonomethylanilin, 2-chlor-n-methylaniline, 2-chloro-n-methyl aniline, 2-chloro-n-methyl-aniline, 2-methylamino chlorobenzene, 2-chloranyl-n-methyl-aniline
PubChem CID 136736
IUPAC Name 2-chloro-N-methylaniline
SMILES CNC1=CC=CC=C1Cl

Specifications

Density 1.15
Boiling Point 218°C
Flash Point 92°C (197°F)
Refractive Index 1.58
Quantity 1 g
UN Number UN2810
Beilstein 2802444
Solubility Information Not miscible or difficult to mix.
Formula Weight 141.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methylaniline
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only