missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(hydroxymethyl)acetamide, 98%

Catalog No. AAA1224718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1224718 50 g
AAA1224709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1224718 Supplier Thermo Scientific Chemicals Supplier No. A1224718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2832-19-1 | C3H6ClNO2 | 123.54 g/mol

An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.

Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 2832-19-1
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.54
MDL Number MFCD00021961
InChI Key TXNSZCSYBXHETP-UHFFFAOYSA-N
Synonym 2-chloro-n-hydroxymethyl acetamide, n-hydroxymethyl-2-chloroacetamide, acetamide, 2-chloro-n-hydroxymethyl, n-methylolchloroacetamide, grota, chloracetamide-n-metholol, n-methylol-chloracetamide, grotan df-35, caswell no. 192bb, chloracetamide-n-methanol
PubChem CID 62466
IUPAC Name 2-chloro-N-(hydroxymethyl)acetamide
SMILES OCNC(=O)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 50 g
UN Number UN3261
Beilstein 1811765
Sensitivity Hygroscopic
Solubility Information Very soluble in water. Poorly soluble in aliphatic hydrocarbons.
Formula Weight 123.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-N- (hydroxymethyl)acetamide
Hazard Category H302+H312+H332-H314-H317-H351
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 220-598-9
RTECSNumber AB5733000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only