accessibility menu, dialog, popup

UserName

CAS RN 2832-19-1

IUPAC Name: 2-chloro-N-(hydroxymethyl)acetamide
Synonyms: 2-chloro-N-(hydroxymethyl)acetamide
Molecular Weight (g/mol): 123.54
Molecular Formula: C3H6ClNO2
InChi Key: TXNSZCSYBXHETP-UHFFFAOYSA-N
SMILES: OCNC(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results