missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide 98.0+%, TCI America™

Catalog No. c32415g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C32415G 5 g
C324125G 25 g
2 options

Catalog No. C32415G

Supplier: TCI America C32415G

Only null left

Chemical Identifiers

CAS 133627-46-0
Molecular Formula C12H9Cl2N3O
Molecular Weight (g/mol) 282.124
MDL Number MFCD09031256
InChI Key UVCHGYJPZGYMSW-UHFFFAOYSA-N
Synonym 2-Chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide
PubChem CID 10423868
IUPAC Name 2-chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide
SMILES CC1=C(C(=NC=C1)Cl)NC(=O)C2=C(N=CC=C2)Cl

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-N-(2-chloro-4-methyl-3-pyridyl)nicotinamide