accessibility menu, dialog, popup

UserName

CAS RN 133627-46-0

IUPAC Name: 2-chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide
Synonyms: 2-chloro-N-(2-chloro-4-methylpyridin-3-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 282.12
Molecular Formula: C12H9Cl2N3O
InChi Key: UVCHGYJPZGYMSW-UHFFFAOYSA-N
SMILES: CC1=C(NC(=O)C2=C(Cl)N=CC=C2)C(Cl)=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results