missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-m-xylene, 98%

Catalog No. AAA1786714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1786714 25 g
AAA1786722 100 g
AAA1786736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1786714 Supplier Thermo Scientific Chemicals Supplier No. A1786714

CAS: 6781-98-2 | C8H9Cl | 140.61 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 6781-98-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000565
InChI Key VDXLAYAQGYCQEO-UHFFFAOYSA-N
Synonym 2-chloro-m-xylene, 2,6-dimethylchlorobenzene, m-xylene, 2-chloro, benzene, 2-chloro-1,3-dimethyl, 2,6-dimethyl chlorobenzene, 2,6-dimethylphenyl chloride, 1-chloro-2,6-dimethylbenzene, 2,6-dimethylchlorobenzene;2-chloro-m-xylene, 2-chloro-meta-xylene, pubchem3643
PubChem CID 32885
IUPAC Name 2-chloro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Cl

Specifications

Melting Point -36°C
Density 1.061
Quantity 25 g
Refractive Index 1.527
Beilstein 2040954
Boiling Point 182°C to 183°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 2-Chloro-m-xylene
Hazard Category H227-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 229-843-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only