missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-L-phenylalanine, 98+%

Catalog No. AAH27135MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27135MD 250 mg
AAH2713503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27135MD Supplier Thermo Scientific Chemicals Supplier No. H27135MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103616-89-3 | C9H10ClNO2 | 199.63 g/mol

The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 103616-89-3
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00077921
InChI Key CVZZNRXMDCOHBG-QMMMGPOBSA-N
Synonym 2-chloro-l-phenylalanine, l-2-chlorophenylalanine, s-2-amino-3-2-chlorophenyl propanoic acid, h-phe 2-cl-oh, 2s-2-amino-3-2-chlorophenyl propanoic acid, phenylalanine, 2-chloro, l-2-chlorophe, l-2-chloro phenylalanine, s-2-chlorophenylalanine, ksc491q0l
PubChem CID 2761491
IUPAC Name (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
SMILES N[C@@H](CC1=C(Cl)C=CC=C1)C(O)=O

Specifications

Melting Point 196°C to 199°C
Solubility Information Insoluble in water.
Quantity 250 mg
Formula Weight 199.64
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only