accessibility menu, dialog, popup

UserName

CAS RN 103616-89-3

IUPAC Name: (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
Synonyms: 2-chloro-L-phenylalanine
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: CVZZNRXMDCOHBG-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=C(Cl)C=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)