missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-beta-nitrostyrene, 98%

Catalog No. AAA1488214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1488214 25 g
AAA1488206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1488214 Supplier Thermo Scientific Chemicals Supplier No. A1488214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3156-34-1 | C8H6ClNO2 | 183.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3156-34-1
Molecular Formula C8H6ClNO2
Molecular Weight (g/mol) 183.591
MDL Number MFCD00024820
InChI Key QHKJTRDWAZGBLR-AATRIKPKSA-N
Synonym 1-chloro-2-2-nitrovinyl benzene, 2-chloro-omega-nitrostyrene, 2-chloro-b-nitrostyrene, e-1-chloro-2-2-nitrovinyl benzene, 1-chloro-2-e-2-nitrovinyl benzene, trans-2-chloro-beta-nitrostyrene, 2-chloro-beta-nitrostyrene, styrene, o-chloro-.beta.-nitro, benzene,1-chloro-2-2-nitroethenyl, 1-chloro-2-e-2-nitroethenyl benzene
PubChem CID 5369313
IUPAC Name 1-chloro-2-[(E)-2-nitroethenyl]benzene
SMILES C1=CC=C(C(=C1)C=C[N+](=O)[O-])Cl

Specifications

Melting Point 46°C to 49°C
Quantity 25 g
Beilstein 2328095
Formula Weight 183.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-beta-nitrostyrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only