missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-9H-carbazole 98.0+%, TCI America™

Catalog No. c33621g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33621G 1 g
C33625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33621G Supplier TCI America Supplier No. C33621G
Only null left

Chemical Identifiers

CAS 10537-08-3
Molecular Formula C12H8ClN
Molecular Weight (g/mol) 201.65
MDL Number MFCD01444890
InChI Key LOQQFCPPDBFFSO-UHFFFAOYSA-N
PubChem CID 2752388
IUPAC Name 2-chloro-9H-carbazole
SMILES ClC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-9H-carbazole