accessibility menu, dialog, popup

UserName

CAS RN 10537-08-3

IUPAC Name: 2-chloro-9H-carbazole
Synonyms: 2-chloro-9H-carbazole
Molecular Weight (g/mol): 201.65
Molecular Formula: C12H8ClN
InChi Key: LOQQFCPPDBFFSO-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(C=C1)C1=C(N2)C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results