missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrotoluene 99.0+%, TCI America™

Catalog No. C023025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023025G 25 g
C0230500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023025G Supplier TCI America Supplier No. C023025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83-42-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007205
InChI Key XCSNRORTQRKCHB-UHFFFAOYSA-N
Synonym 2-chloro-6-nitrotoluene, 6-chloro-2-nitrotoluene, benzene, 1-chloro-2-methyl-3-nitro, toluene, 2-chloro-6-nitro, 3-chloro-2-methylnitrobenzene, caswell no. 202, unii-wez013857x, epa pesticide chemical code 019001, chloro-o-nitrotoluene, pubchem14277
PubChem CID 6740
IUPAC Name 1-chloro-2-methyl-3-nitrobenzene
SMILES CC1=C(Cl)C=CC=C1[N+]([O-])=O

Specifications

Color White-Yellow
Boiling Point 241°C
Quantity 25 g
UN Number 3457
Formula Weight 171.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-6-nitrotoluene
EINECSNumber (3)-0449
RTECSNumber XS9130000
TSCA Yes