missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzoic acid, 99%

Catalog No. AAL1705903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1705903 1 g
AAL1705906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1705903 Supplier Thermo Scientific Chemicals Supplier No. L1705903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5344-49-0 | C7H4ClNO4 | 201.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5344-49-0
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00100425
InChI Key JYHOMEFOTKWQPN-UHFFFAOYSA-N
Synonym 2-chloro-6-nitrobenzoic acid, 2-chloro-6-nitro-benzoic acid, 6-chloro-2-nitrobenzoic acid, benzoic acid, 2-chloro-6-nitro, pubchem20037, 6-chloro-2-nitrobenzo, acmc-209l7b, 2-chloro-6-nitrobenzoicacid, 2-carboxy-3-chloronitrobenzene, 2-chloro-6-nitrobenzoic acid #
PubChem CID 79287
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 161°C to 166°C
Odor Odorless
Quantity 1 g
Beilstein 2694949
Formula Weight 201.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-6-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-287-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only