missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylpyridine 98.0+%, TCI America™
Supplier: TCI America C165425G
Specifications
| 2-Chloro-6-methylpyridine | |
| Yellow | |
| C6H6ClN | |
| 25 g | |
| GXZDYRYYNXYPMQ-UHFFFAOYSA-N | |
| 2-chloro-6-methylpyridine | |
| 87601 | |
| ≥98.0% (GC) |
| 18368-63-3 | |
| 88°C | |
| MFCD00006245 | |
| 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine | |
| CC1=CC=CC(Cl)=N1 | |
| 127.57 | |
| 127.57 | |
| Liquid |
Chemical Identifiers
| 18368-63-3 | |
| 127.57 | |
| GXZDYRYYNXYPMQ-UHFFFAOYSA-N | |
| 87601 | |
| CC1=CC=CC(Cl)=N1 |
| C6H6ClN | |
| MFCD00006245 | |
| 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine | |
| 2-chloro-6-methylpyridine |
Safety and Handling
TSCA : No