missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylpyridine, 98%

CAS: 18368-63-3 | C6H6ClN | 127.57 g/mol

Supplier:  Thermo Scientific Chemicals B2017406

 View more versions of this product

Catalog No. AAB2017406


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-6-methylpyridine
1.167
73°C (163°F)
1.5265
5 g
107187
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
2-chloro-6-methylpyridine
87601
98%
18368-63-3
64°C to 68°C (10 mmHg)
C6H6ClN
MFCD00006245
UN2810
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
CC1=CC=CC(Cl)=N1
127.57
127.57

Chemical Identifiers

18368-63-3
127.57
GXZDYRYYNXYPMQ-UHFFFAOYSA-N
87601
CC1=CC=CC(Cl)=N1
C6H6ClN
MFCD00006245
6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridine
2-chloro-6-methylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 242-241-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only