missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylbenzaldehyde, 98%

Catalog No. AAH2602303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2602303 1 g
AAH2602306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2602303 Supplier Thermo Scientific Chemicals Supplier No. H2602303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1194-64-5 | C8H7ClO | 154.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1194-64-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD01934409
InChI Key CCYFXIJPJFSTSU-UHFFFAOYSA-N
PubChem CID 12467050
IUPAC Name 2-chloro-6-methylbenzaldehyde
SMILES CC1=C(C(=CC=C1)Cl)C=O

Specifications

Melting Point 36°C to 38°C
Boiling Point 74°C (0.4 mmHg)
Flash Point 99°C (210°F)
Quantity 1 g
Sensitivity Air Sensitive
Synonym 3-chloro tolualdehyde, pubchem19866, 3-chloro-2-formyltoluene, 2-chloro-6-methylbenzadehyde, 6-chloro-2-methylbenzaldehyde, 2-chloro-6-methyl-benzaldehyde, benzaldehyde,2-chloro-6-methyl, 2-chloranyl-6-methyl-benzaldehyde, benzaldehyde, 2-chloro-6-methyl, 2-chloro-6-methylbenzaldehyde
Formula Weight 154.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only