missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline 98.0+%, TCI America™

Catalog No. p-7117657
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19595G 5 g
A195925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19595G Supplier TCI America Supplier No. A19595G

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007679
InChI Key WFNLHDJJZSJARK-UHFFFAOYSA-N
Synonym 6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
PubChem CID 6897
IUPAC Name 2-chloro-6-methylaniline
SMILES CC1=CC=CC(Cl)=C1N

Specifications

Melting Point 12°C
Boiling Point 215°C
Quantity 5 g
UN Number 3429
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-methylaniline
EINECSNumber (9)-0073&(3)-0265
RTECSNumber XU5100000
TSCA Yes