missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylaniline, 98%

Catalog No. AAL0262914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0262914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0262914 Supplier Thermo Scientific Chemicals Supplier No. L0262914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-63-8 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-63-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007679
InChI Key WFNLHDJJZSJARK-UHFFFAOYSA-N
Synonym 6-chloro-2-methylaniline, 6-chloro-o-toluidine, 2-amino-3-chlorotoluene, benzenamine, 2-chloro-6-methyl, 3-chloro-2-aminotoluene, 6-chloro-2-toluidine, 2-methyl-6-chloroaniline, o-toluidine, 6-chloro, 2-chloro-6-methyl-aniline, 2-chloro-6-methylaninile
PubChem CID 6897
IUPAC Name 2-chloro-6-methylaniline
SMILES CC1=CC=CC(Cl)=C1N

Specifications

Melting Point 2°C
Density 1.174
Boiling Point 215°C
Flash Point 98°C (208°F)
Refractive Index 1.5775
Quantity 25 g
UN Number UN3429
Beilstein 774624
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, LIQUID
EINECSNumber 201-759-2
RTECSNumber XU5100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only