missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7125547
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
C25941G 1 g
C25945G 5 g
C259425G 25 g
3 options

Catalog No. C25941G

Supplier: TCI America C25941G

Only null left

Chemical Identifiers

CAS 56057-19-3
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD03085820
InChI Key UIEVSGOVFXWCIK-UHFFFAOYSA-N
Synonym 6-chloro-5-nitro-2-picoline, 2-chloro-3-nitro-6-methylpyridine, 2-chloro-3-nitro-6-picoline, 2-chloro-3-nitro-6-methyl pyridine, 2-chloro-6-methyl-3-nitro-pyridine, pyridine, 2-chloro-6-methyl-3-nitro, 2-chloro-6-methyl-3-nitropyridine?, pubchem6175, acmc-209lqq
PubChem CID 1268230
IUPAC Name 2-chloro-6-methyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 72°C
Color Yellow
Quantity 1 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methyl-3-nitropyridine