missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxytoluene 96.0+%, TCI America™

Catalog No. p-7125073
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15851G 1 g
C15855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15851G Supplier TCI America Supplier No. C15851G

Chemical Identifiers

CAS 3260-88-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00070772
InChI Key LTVRGAWOEOKGJZ-UHFFFAOYSA-N
Synonym 3-chloro-2-methylanisole, 2-methyl-3-chloroanisole, 2-chloro-6-methoxytoluene, benzene, 1-chloro-3-methoxy-2-methyl, 2-methoxy-6-chlorotoluene, 1-chloro-3-methoxy-2-methyl-benzene, pubchem2664, acmc-1cq3o, ltvrgawoeokgjz-uhfffaoysa
PubChem CID 76749
IUPAC Name 1-chloro-3-methoxy-2-methylbenzene
SMILES COC1=CC=CC(Cl)=C1C

Specifications

Color Colorless
Boiling Point 207°C
Quantity 1 g
Formula Weight 156.61
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-methoxytoluene