missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxypyridine 98.0+%, TCI America™

Catalog No. C01975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01975G 5 g
C019725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01975G Supplier TCI America Supplier No. C01975G

Chemical Identifiers

CAS 17228-64-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00006265
InChI Key VAVGOGHLNAJECD-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-6-methoxy, 2-methoxy-6-chloropyridine, 6-chloro-2-methoxypyridine, 2-chloro-6-methoxy-pyridine, 2-chlor-6-methoxypyridin, pubchem13194, acmc-1cffb, 6-methoxy-2-chloropyridine, 2-chloro-6-methoxy pyridine, 2-chloro-6-methyloxy pyridine
PubChem CID 87009
IUPAC Name 2-chloro-6-methoxypyridine
SMILES COC1=NC(=CC=C1)Cl

Specifications

Color Colorless
Boiling Point 184°C
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-methoxypyridine